Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam L-Methionine, Essential Amino Acid, >99% Purity, 25 g

Catalog No. 502704801
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4801 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4801 Supplier Abcam Supplier No. AB14624825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Essential amino acid with >99% purity, serving as a versatile methyl-group donor and chelating agent.

  • Molecular Weight: 149.21 g/mol
  • Purity: >99%
  • Composition: 100% L-Methionine
  • CAS Number: 63-68-3
  • Applications: Essential amino acid for human development, hepatoprotectant, acetaminophen poisoning antidote, urinary acidifier, heavy metal chelating agent, and in vivo methyl-group donor

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Form Powder
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Formulation 100% L-Methionine
Quantity 25 g
For Use With (Application) Hepatoprotection, Acetaminophen poisoning treatment, Urinary acidification, Heavy metal chelation, In vivo methyl-group donation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.