Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Methionine sulfoxide

Catalog No. AAJ6287306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6287306 5 g
AAJ6287303 1 g
AAJ6287314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6287306 Supplier Thermo Scientific Chemicals Supplier No. J6287306
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3226-65-1 | C5H11NO3S | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3226-65-1
Molecular Formula C5H11NO3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00063093
InChI Key QEFRNWWLZKMPFJ-YGVKFDHGSA-N
Synonym l-methionine sulfoxide, h-met o-oh, methionine s-oxide, methionine sulfoxide, 2s-2-amino-4-methylsulfinyl butanoic acid, l-methionine s-oxide, 2s-2-amino-4-methylsulfinylbutanoic acid, l-2-amino-4-methylsulfinyl butanoic acid, l-methionine-dl-sulfoxide, 2s-2-azanyl-4-methylsulfinyl-butanoic acid
PubChem CID 158980
ChEBI CHEBI:17016
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](N)C(O)=O

Specifications

Color White
Beilstein 4129939
Quantity 5 g
Formula Weight 165.21
Physical Form Powder
Chemical Name or Material L-Methionine sulfoxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-758-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.