Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam L-Mimosine, Non-Protein Amino Acid, >98% Purity, Solid, 25 mg

Catalog No. 502705069
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5069 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5069 Supplier Abcam Supplier No. AB14586125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toxic non-protein amino acid that arrests the cell cycle by chelating iron and inhibiting deoxyribonucleotide metabolism.

  • Purity: >98%
  • Molecular Weight: 198.18 g/mol
  • Form: Solid
  • Chemical Class: Toxic non-protein amino acid
  • Biological Activity: Arrests cell cycle at G1 phase (PC-3 cells) and S phase (LNCaP cells); blocks cell cycle progression by iron chelation in MDA-MB-453 human breast cancer cells; inhibits deoxyribonucleotide metabolism to arrest DNA synthesis; acts as a prolyl hydroxylase (PHD) inhibitor to stimulate VEGF production and increase proangiogenic capacity in dental pulp-derived cells
  • Composition: 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Buffer 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Shipping Condition Ambient
Target Molecule Prolyl Hydroxylase, Iron, DNA
Form Solid
Formulation 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Quantity 25 mg
For Use With (Application) Cell cycle arrest, DNA synthesis inhibition, Prolyl hydroxylase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.