Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam L-Mimosine, Non-protein Amino Acid, >98% Purity, Solid, 250 mg

Catalog No. 502705070
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5070 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5070 Supplier Abcam Supplier No. AB145861250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toxic non-protein amino acid that arrests the cell cycle by inhibiting deoxyribonucleotide metabolism and chelating iron.

  • Molecular Weight: 198.18 g/mol
  • Purity: >98%
  • Form: Solid
  • Description: Toxic non-protein amino acid originally isolated from Mimosa pudica.
  • Mechanism of Action: Arrests cell cycle at the G1 phase in PC-3 cells and at the S phase in LNCaP cells; blocks cell cycle progression by chelating iron; inhibits DNA synthesis by inhibiting deoxyribonucleotide metabolism; acts as a prolyl hydroxylase (PHD) inhibitor.
  • Storage Temperature: Store at -20°C.
  • Chemical Formula: (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Prolyl Hydroxylase
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Content And Storage Store at -20°C.
Form Solid
Formulation 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Quantity 250 mg
For Use With (Application) Cell cycle arrest, DNA synthesis inhibition, Prolyl hydroxylase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.