Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam L-Mimosine, Non-Protein Amino Acid, Solid, >98% Purity, 100 mg

Catalog No. 502705071
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5071 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5071 Supplier Abcam Supplier No. AB145861100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toxic non-protein amino acid that arrests cell cycle progression by chelating iron.

  • Chemical Name: (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
  • Purity: >98%
  • Molecular Weight: 198.18 g/mol
  • Form: Solid
  • CAS Number: 500-44-7
  • Biological Activity: Toxic non-protein amino acid that arrests cell cycle (G1 phase in PC-3 cells, S phase in LNCaP cells), blocks cell cycle progression by iron chelation, inhibits DNA synthesis, and acts as a prolyl hydroxylase (PHD) inhibitor.
  • Storage: Store at -20°C.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Prolyl Hydroxylase
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Form Solid
Buffer 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Content And Storage Store at -20°C.
Formulation 100% (S)-α-Amino-3-hydroxy-4-oxo-1(4H)-pyridylpropionic acid
Quantity 100 mg
For Use With (Application) Cell cycle research, DNA synthesis inhibition, Prolyl hydroxylase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.