Learn More
Cayman Chemical L-NAME (hydrochloride), 51298-62-5, 5 g
SDP
Supplier:  Cayman Chemical 802105
Encompass_Preferred
Molecular Formula C7H15N5O4 • HCl, Purity ≥99%, Formula Weight 269.7, Synonyms: L-NG-Nitroarginine methyl ester, N(G)-Nitro-L-arginine methyl ester; A methyl ester prodrug form of the NOS inhibitor L-NNA; exhibits Ki values of 15 nM, 39 nM, and 4.4 M for nNOS (bovine), eNOS (human), and iNOS (mouse), respectively.
Catalog No. NC0104875
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.