Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Ornithine Hydrochloride, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611376 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-376 5 kg Poly Pail
18-611-374 25 g Poly Bottle
18-611-373 100 g Poly Bottle
18-611-377 500 g Poly Bottle
18-611-372 1 kg Poly Bottle
18-611-375 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-376 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. OR1305KGBL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.003
Packaging Poly Pail
Quantity 5 kg
Specific Rotation +23° to +25.0°
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.