Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylephrine 98.0+%, TCI America™
SDP

Catalog No. P039525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P039525G 25 g
P03955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P039525G Supplier TCI America Supplier No. P039525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59-42-7
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00044749
InChI Key SONNWYBIRXJNDC-VIFPVBQESA-N
Synonym (R)-(-)-Phenylephrine, (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol
PubChem CID 6041
ChEBI CHEBI:8093
IUPAC Name 3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol
SMILES CNCC(C1=CC(=CC=C1)O)O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 25 g
Formula Weight 167.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Phenylephrine
RTECSNumber DO7175000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.