Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Prolinamide 97.0+%, TCI America™
SDP

Catalog No. P138225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P138225G 25 g
P13821G 1 g
P13825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P138225G Supplier TCI America Supplier No. P138225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
IUPAC Name (2S)-2-carbamoylpyrrolidin-1-ium
SMILES NC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 25 g
Formula Weight 114.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Prolinamide
EINECSNumber (5)-6773
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.