Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™

Catalog No. ICN19472825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472825 25 g
ICN19472880 100 g
02-194-72891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19472825 Supplier MP Biomedicals, Inc Supplier No. 0219472825
Add to Cart
Add to Cart

L-Proline, Cell Culture Reagent, ≥99% is used in cell culture media and as a component of MEM non-essential amino acids solution.

  • L-Proline is used in cell culture media.
  • L-Proline is also a component of MEM non-essential amino acids solution.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-85-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00064318
InChI Key ONIBWKKTOPOVIA-UHFFFAOYNA-N
Synonym l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
PubChem CID 145742
ChEBI CHEBI:17203
IUPAC Name pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1

Specifications

Melting Point 221°C
Solubility Information Soluble in water, 162,000 mg/L at 25°C
Density 1.064 g/mL at 24°C
Quantity 25 g
Optical Rotation -82° to -85° (c=1.6, water)
Formula Weight 115.1
Percent Purity ≥99%
Grade Cell Culture
Physical Form Powder
pKa 1.95 (COOH), 10.64 (NH2)
Chemical Name or Material L-Proline, Hydroxy-L-proline free
Show More Show Less
Recommended Storage Room temperature

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.