Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ L-Selectin/CD62L Antibody Pair [HRP]
SDP

Catalog No. NB009893 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009893 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009893 Supplier Novus Biologicals™ Supplier No. NBP27942715PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 12.5 pg/mL
Assay Sensitivity 12.5-800 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse L-Selectin/CD62L.
Target Species Mouse
For Use With (Application) Sandwich ELISA
Gene Alias CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L
Gene ID (Entrez) 6402
Gene Symbol SELL
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.25 mg/mL rabbit anti-L-Selectin/CD62L polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse L-Selectin/CD62L monoclonal antibody.
  • Standard: Each vial contains 32 ng of recombinant mouse L-Selectin/CD62L.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target L-Selectin/CD62L
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 800 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.