Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-selectin Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7211726 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5354 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5354 Supplier Proteintech Group Inc Supplier No. 264771AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The human CD62L is a 74-95 kD glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.

Specifications

Antigen L-selectin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SELL
Gene Accession No. P14151
Gene Alias CD62L, gp90 MEL, hLHRc, LAM 1, LAM1, LECAM1, Leu 8, Leukocyte adhesion molecule 1, LNHR, LSEL, L-selectin, Lyam 1, LYAM1, Lymph node homing receptor, PLNHR, selectin L, SELL, TQ1
Gene Symbols SELL
Host Species Rabbit
Immunogen L-selectin Fusion Protein Ag24063
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6402
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.