Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serine Benzyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. S05505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05505G Supplier TCI America Supplier No. S05505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60022-62-0
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00038955
InChI Key MGZWCDQAKCHOBX-UHFFFAOYNA-N
Synonym l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride
PubChem CID 12930233
IUPAC Name benzyl 2-amino-3-hydroxypropanoate hydrochloride
SMILES Cl.NC(CO)C(=O)OCC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 231.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Serine Benzyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.