Learn More
Abcam L-Serine, Non-essential Amino Acid, 25 g
Description
- Molecular Weight: 105.09 g/mol
- Composition: 100% L-Serine
- Biological Role: Non-essential amino acid used as a precursor for amino acids including glycine and cysteine, metabolites such as sphingolipids and folate, and in the biosynthesis of purines and pyrimidines
- Functional Role: Plays a role in enzyme catalytic function and is essential for cell proliferation and central nervous system functions
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | L-Serine |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% L-Serine |
| Formulation | 100% L-Serine |
| Quantity | 25 g |
| For Use With (Application) | Bio-synthesis of purines and pyrimidines, cell proliferation, central nervous system research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.