Learn More
United Scientific™ L-Shaped Spreader
SDPSCI_ED
Catalog No. S111019 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
S111019 100 Pk.
1 options
Catalog No. S111019 Supplier United Scientific™ Supplier No. UNLSPREADPK/100
Add to Cart
Add to Cart
L-shaped spreaders are a convenient way of spreading samples on petri dishes.
  • Featuring a smooth and rounded edged to prevent gouging or scraping while spreading samples
Specifications
Disposable Single-use
Shape L Spreader
Quantity 100 Pk.
For Use With (Equipment) General Lab Use
Material Polypropylene
Product Type Spreader
Sterility Sterile
For Use With (Application) Spreading samples on an agar dish