Description
L-shaped spreaders
L-Shaped Plastic Spreaders
- For spreading liquid samples
- Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
- Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
- Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Specifications
| Quantity | 5/Peel Pk., 1000 Spreaders/Box |
| Product Type | Cell Spreader |
| For Use With (Equipment) | Culture Plates |
| Shape | L-Shape |
| Sterility | Sterile |
| Material | Plastic |
| For Use With (Application) | Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |