Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Sorbitol 97.0+%, TCI America™
SDP

Catalog No. S0388100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0388100MG 100 mg
S03881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0388100MG Supplier TCI America Supplier No. S0388100MG

Chemical Identifiers

CAS 6706-59-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00191490
InChI Key FBPFZTCFMRRESA-FSIIMWSLSA-N
PubChem CID 82170
ChEBI CHEBI:28789
IUPAC Name (2R,3S,4S,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 91°C
Color White
pH 5 to 6 (1% H2O soln.)
Quantity 100 mg
Formula Weight 182.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material L-Sorbitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.