Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Taurine USP/JP, Ajinomoto

Catalog No. 502946102
Encompass
Change view
Click to view available options
Quantity:
20 kg
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-294-6102 50 kg
50-294-5965 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-294-6102 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A20069

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Weight (g/mol) 125.15

Specifications

Quantity 50 kg
Percent Purity 98.5 - 101.0%
Grade USP
Physical Form Fine Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.