Learn More
U.S. Pharmacopeia L-Threonine, 72-19-5, MFCD00064270, 200mg
Supplier:  U.S. Pharmacopeia 1667202
Encompass
Molecular formula C4H9NO3, Molecular weight 119.12, Melting point/freezing point 491 - 494.6 °F (255 - 257 °C) (decomposes), Synonym: Threonine
Catalog No. NC1606469
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.