Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Thymidine

Catalog No. AC464550050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464550050 5 g
AC464550010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464550050 Supplier Thermo Scientific Chemicals Supplier No. 464550050
Add to Cart

missing translation for 'validMainframeMsg'

242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Format Crystalline Powder
Molecular Weight (g/mol) 242.23
Product Type L-Thymidine
Solution Clarity (1% aq. soln.) Clear colorless solution
Source synthetic source
Content And Storage Refrigerator +4°C
Shipping Condition normal conditions
Purity ≥98% by HPLC
Quantity 5 g
Color White to almost white
ShelfLife 5 years

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.