Learn More
Cayman Chemical LoratdIn-d5 5mg
SDP
Supplier:  Cayman Chemical 250405 MG
Encompass_Preferred
A prodrug form of LBQ657 increases plasma levels of ANF in concious rats and anesthetized dogs at 10 and 30 mg/kg respectively increases ANF-induced urinary sodium excretion in anesthetized rats at 30 mg/kg
Catalog No. 50-293-3580
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.