Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L-Thyroxine T4 Monoclonal Antibody (T4YCH)
GREENER_CHOICE

Catalog No. PIMA514716 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514716 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514716 Supplier Invitrogen™ Supplier No. MA514716
  • $672.00
    $481.65 / Each of 1

    Save $190.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA5-14716 targets L-Thyroxine (T4) in ELISA, Western Blot, IP, and RIA applications and shows reactivity with Chemical samples. Product MA514716 is a smaller package size of MIT0501 (formerly sold as a Seradyn product).

Levothyroxine, also L-thyroxine, synthetic T4, or 3,5,3',5'-tetraiodo-L-thyronine, is a synthetic form of thyroxine (thyroid hormone), used as a hormone replacement for patients with thyroid problems. The natural hormone is chemically in the chiral L-form, as is the pharmaceutical agent. Dextrothyroxine (D-thyroxine) briefly saw research as an anticholesterol agent but was pulled due to cardiac side-effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L-Thyroxine T4
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone T4YCH
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Alias C15H11I4NO4; Levothyroxine; L-thyroxine; Synthroid; Thyroxine
Host Species Mouse
Immunogen Thyroxine-BSA.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.