Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam L-trans-Pyrrolidine-2,4-dicarboxylic acid, EAAT2, EAAT4 and EAAT5 inhibitor, 10 mg

Catalog No. 502704942
Encompass_Preferred
Catalog No. 50-270-4942 Supplier Abcam Supplier No. AB14613110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-trans-Pyrrolidine-2, inhibitor.

  • Class: Inhibitor
  • Form: Solid
  • Purity: >99%
  • Molecular Weight: 159.14 g/mol

Specifications

Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.