Learn More
Abcam L-Tryptophan, >99% Purity, 50 g
Description
- Molecular weight: 204.2 g/mol
- Purity: >99%
- Storage buffer: Constituents: 100% L-Tryptophan
- Key biological roles: Necessary for normal growth in infants and for nitrogen balance in adults; precursor to niacin, indole alkaloids, serotonin, and melatonin
- Applications: Used as an antidepressant, anxiolytic, and sleep aid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% L-Tryptophan |
| Formulation | 100% L-Tryptophan |
| Quantity | 50 g |
| For Use With (Application) | Antidepressant, anxiolytic, sleep aid, precursor to niacin, indole alkaloids, serotonin and melatonin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.