Learn More
Ampco Safety Tools L-Type Socket Wrench (Metric)
SDP
Catalog No. 19174348 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Drive Size:
8 mm
9 mm
10 mm
11 mm
13 mm
14 mm
15 mm
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
32 mm
33 mm
35 mm
36 mm
37 mm
41 mm
42 mm
46 mm
47 mm
50 mm
52 mm
54 mm
55 mm
56 mm
60 mm
63 mm
65 mm
67 mm
70 mm
71 mm
75 mm
77 mm
80 mm
82 mm
46 product options available for selection
Product selection table with 46 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Drive Size
19-174-348 26 mm
19-174-299 10 mm
19-174-300 11 mm
19-174-310 13 mm
19-174-316 14 mm
19-174-317 15 mm
19-174-321 16 mm
19-174-322 17 mm
19-174-325 18 mm
19-174-326 19 mm
19-174-330 20 mm
19-174-331 21 mm
19-174-338 22 mm
19-174-339 23 mm
19-174-343 24 mm
19-174-344 25 mm
19-174-349 27 mm
19-174-351 28 mm
19-174-352 29 mm
19-174-357 30 mm
19-174-361 32 mm
19-174-362 33 mm
19-174-364 35 mm
19-174-365 36 mm
19-174-366 37 mm
19-174-368 41 mm
19-174-369 42 mm
19-174-370 46 mm
19-174-371 47 mm
19-174-374 50 mm
19-174-375 52 mm
19-174-376 54 mm
19-174-377 55 mm
19-174-378 56 mm
19-174-380 60 mm
19-174-381 63 mm
19-174-382 65 mm
19-174-383 67 mm
19-174-386 70 mm
19-174-387 71 mm
19-174-388 75 mm
19-174-389 77 mm
19-174-390 8 mm
19-174-391 80 mm
19-174-392 82 mm
19-174-393 9 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 46 options available.
46 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-348 Supplier Ampco Safety Tools Supplier No. SL26
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ L-Type Socket Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type L-Type Socket Wrench
Drive Size 26 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.