Learn More
Cayman Chemical 3ACtyldeoy NIvalenl13C17 1.2mL
SDP
Supplier:  Cayman Chemical 312581.2 ML
Encompass_Preferred
An inhibitor of PDK1 (IC50 6 nM) inhibits PDK1-induced Akt2 activation in a cell-free assay (IC50 20 nM)
Catalog No. 50-298-9210
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.