Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine-d11, Thermo Scientific™

Catalog No. AC233391000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC233391000 100 mg
AC233395000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233391000 Supplier Thermo Scientific Chemicals Supplier No. 233391000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39748-77-1
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 192.258
InChI Key OUYCCCASQSFEME-VQWUOSHWSA-N
Synonym l-tyrosine-d11
PubChem CID 90471999
IUPAC Name deuterio (2S)-2,3,3-trideuterio-2-(dideuterioamino)-3-(2,3,5,6-tetradeuterio-4-deuteriooxyphenyl)propanoate
SMILES C1=CC(=CC=C1CC(C(=O)O)N)O

Specifications

Quantity 100 mg
Linear Formula 4(DO)C6D4CD2CD(ND2)CO2D
Formula Weight 192.28
Chemical Name or Material L-Tyrosine-d11
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.