Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine disodium salt, 98%

Catalog No. AC426415000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426415000 500 g Glass Bottle
AC426410250 25 g Glass Bottle
AC426411000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC426415000 Supplier Thermo Scientific Chemicals Supplier No. 426415000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 69847-45-6 | C9H9NNa2O3 | 225.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69847-45-6
Molecular Formula C9H9NNa2O3
Molecular Weight (g/mol) 225.16
MDL Number MFCD00037203
InChI Key ASIYFCYUCMQNGK-RIYJSUBBNA-L
Synonym sodium s-2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 66602062
SMILES [Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Melting Point >300.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula NaOC6H4CH2CH(NH2)CO2Na
Specific Rotation − 12.90
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid;sodium
Quantity 500 g
Formula Weight 225.15
Percent Purity 98%
Chemical Name or Material L-Tyrosine disodium salt
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.