Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Valinol 97.0+%, TCI America™
SDP

Catalog No. V005825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V005825G 25 g
V00585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V005825G Supplier TCI America Supplier No. V005825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2026-48-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064296
InChI Key NWYYWIJOWOLJNR-RXMQYKEDSA-N
Synonym l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol
PubChem CID 640993
IUPAC Name (2S)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@H](N)CO

Specifications

Color White-Yellow
Boiling Point 88°C
Quantity 25 g
Formula Weight 103.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material L-Valinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.