Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Active Motif L3MBTL1 Polyclonal Antibody

Catalog No. SPN39183 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPN-39183 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPN-39183

Rabbit Polyclonal Antibody

L3MBTL1 Polyclonal Antibody for Western Blot

L3MBTL1 is the homolog of a protein identified in Drosophila as a suppressor of malignant transformation of neuroblasts and ganglion-mother cells in the optic centers of the brain. L3MBTL1 is localized to condensed chromosomes in mitotic cells and overexpression of this protein in a glioma cell line results in improper nuclear segregation and cytokinesis producing multinucleated cells. L3MBTL1 represents a polycomb group gene the encoded protein functions to regulate gene activity, likely via chromatin modification. Polycomb group protein specifically recognizes and binds mono- and dimethyllysine residues on target proteins, thereby acting as a reader of a network of post-translational modifications. Alternatively spliced transcript variants encoding different isoforms of the L3MBTL1 gene have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L3MBTL1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide
Gene L3MBTL1
Gene Accession No. Q9Y468
Gene Alias C630004G01; dJ138B7.3; H-l(3)mbt; H-l(3)mbt protein; Kiaa0681; L(3)mbt protein homolog; l(3)mbt-like (Drosophila); l(3)mbt-like 1; l(3)mbt-like 1 (Drosophila); L3mbt; L3MBTL; L3MBTL histone methyl-lysine binding protein 1; L3MBTL1; L3MBTL1, histone methyl-lysine binding protein; lethal (3) malignant brain tumor l(3); lethal(3)malignant brain tumor-like protein 1; lethal(3)malignant brain tumor-like protein 1; serine/threonine-protein kinase Sgk2; LOC100070683; mKIAA0681; RGD1307316; RP1-138B7.1; ZC2HC3
Gene Symbols L3MBTL1
Host Species Rabbit
Immunogen This L3MBTL1 antibody was raised against a His-Tagged fusion protein corresponding to amino acids 1-215 of human L3MBTL1.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26013
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.