Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L3MBTL2 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188510
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4058 10 μL
50-199-4057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4058 Supplier Active Motif Supplier No. 39570
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L3MBTL2 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.

Specifications

Antigen L3MBTL2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene L3MBTL2
Gene Accession No. Q969R5
Gene Alias 4732493N06Rik; BOS_6190; H-l(3)mbt-l; H-l(3)mbt-like protein 2; l(3)mbt-like 2; l(3)mbt-like 2 (Drosophila); l(3)mbt-like 2 protein; L(3)mbt-like protein 2; L3MBT; L3mbtl2; L3MBTL2 polycomb repressive complex 1 subunit; L3MBTL2, polycomb repressive complex 1 subunit; lethal(3)malignant brain tumor-like 2 protein; Lethal(3)malignant brain tumor-like protein 2; lmbt-like 2 (Drosophila) isoform 1; lmbt-like 2 (Drosophila) isoform 2; M4mbt
Gene Symbols L3MBTL2
Host Species Rabbit
Immunogen This L3MBTL2 antibody was raised against a His-Tagged fusion protein containing amino acids 1-230 of human L3MBTL2.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83746
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.