Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Lorazepam-d4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco L9021ML

Catalog No. 01-001-559


Only null left
Add to Cart

Description

Description

A stable-labeled internal standard suitable for quantitation of lorazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis. Lorazepam, also known by trade names Ativan™ or Orfidal, is a benzodiazepine often prescribed for treatment of anxiety disorders.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

75-05-8, 84344-15-0
MFCD00673284
100 μg/mL in acetonitrile
Ampule
C15D4H6Cl2N2O2
UN 1648 3 / PGII
325.18
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-835-2

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation