Learn More
Siemens Healthineers LA 1 Screening Reagent
Catalog No. 23044726 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 2 mL
Catalog No. Quantity
23-044-726 10 x 2 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-726 Supplier Siemens Healthineers Supplier No. 10461887
Add to Cart
Add to Cart
This item is not returnable. View return policy
Simplified DRVV reagent to screen for the presence of Lupus Anticoagulants.
LA 1 Screening Reagent contains dilute Russell’s viper venom and low phospholipids for use in the simplified DRVVT as a screening test for lupus anticoagulants. The LA 1 Screening Reagent was designed to be used in conjunction with the LA 2 Confirmation Reagent.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombophilia Testing
Quantity 10 x 2 mL