Learn More
Siemens Healthineers LA 2 Confirmation Reagent
Catalog No. 23044715 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Catalog No. Quantity
23-044-715 10 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-715 Supplier Siemens Healthineers Supplier No. 10458687
Add to Cart
Add to Cart
This item is not returnable. View return policy
Phospholipid-rich DRVV reagent for the specific correction of Lupus Anticoagulants.
A phospholipid-rich DRVVT reagent, the LA 2 Confirmation Reagent is used for the specific correction of lupus anticoagulants. The LA 2 Confirmation Reagent was designed to be used in conjunction with the LA 1 Screening Reagent.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombophilia Testing
Quantity 10 x 1 mL