Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

La Crosse Virus Glycoprotein 1 Antibody (807.31), Alexa Fluor™ 647 - Chimeric, Novus Biologicals™
SDP

Catalog No. NB232915
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232915 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB232915 Supplier R&D Systems Supplier No. NBP320122AF647

Customers require a valid business account associated to their profile to access pricing and ordering options.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Monoclonal Antibody

La Crosse Virus Glycoprotein 1 Monoclonal specifically detects La Crosse Virus Glycoprotein 1 in Virus samples. It is validated for ELISA∣Flow Cytometry∣Immunoprecipitation∣Neutralization∣Western Blot.

Specifications

Antigen La Crosse Virus Glycoprotein 1
Applications ELISA, Flow Cytometry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone 807.31
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Bunyavirus La Crosse Glycoprotein 1, G1 envelope glycoprotein, Glycoprotein C, La Crosse Virus Gc, LCV G1, M polyprotein G1 chain
Host Species Rabbit
Immunogen This antibody was raised by immunising BALB/c mice with La Crosse viruses and the harvested spleens were fused with mouse P3X63Ag8.653 myeloma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956555
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.