Learn More
Braintree Scientific La Pias Ear Tags, Rat, 5 Colors 1-20/ea
SDP
Supplier:  Braintree Scientific 5678520
Encompass
La Pias Ear Tags - Animal Identification Made Easy Specially suited for use with small lab and research animals (mice, rats and other rodents). The light weight easy-to-read tags allow for trouble-free identification. Ear tags are numbered from 1-100 (available up to 999) Available as Stainless Steel (non-colored) or in 5 Once applied, they cannot be easily removed Laser-etched number provides durability Material Numbered Tag: Aluminum Clasp: Stainless Steel Size Mouse: 3.5mm O.D. with 3mm PinRat: 3.5mm O.D. with 5mm Pin **Sold for research purposes only and not for human use**
Catalog No. 50-195-4116
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.