Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab-Aids™ Antacid Study Kit
SCI_ED

Catalog No. S19476
Encompass_Preferred
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S19476 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S19476 Supplier Lab-Aids™ Supplier No. 181
This item is not returnable. View return policy

Includes

Calibrated tubes with caps (24); 60mL Hydrochloric Acid, 0.5M (2); 60mL Sodium Hydroxide, 0.5M (2); 180mL Hydrochloric Acid (fill is 100mL), 0.1M (4); 30mL drop-controlled bottles of Sodium Hydroxide, 0.1M (8); 15mL drop-controlled bottle of Phenolphthalein solution; universal pH indicator strips; 60mL Limewater (2); Chemplates™ (8); rubber tubes (8); student worksheets and guides (30); and an instruction manual with SDS

Requires

Commercial/over-the-counter antacids

Relate science to the real world by reinforcement and application, perform a neutralizing activity and quantitize it, infer which brands of antacids are most effective and determine cost-benefits of various brands.

This kit makes science more interesting and meaningful by dealing with fundamental chemical principles and applying them to products found at home. Most antacids are composed of bases or alkalizing agents. Students use an over-the-counter antacid (e.g. Rolaids, Tums, etc.) brought from home and perform a neutralizing activity to determine how much acid can be neutralized by a particular brand. After conducting the activities, students make comparisons and draw conclusions about the different brands tested.

This kit is complete for 30 students. Note: This kit does not contain antacids; they must be supplied. Accommodates 2 classes, each with 8 groups of 4 students. Includes Teacher guide, student sheets, assessment questions, SDS for all chemicals in kits.

Specifications

For Use With (Application) Use over the counter antacids to determine how much acid can be neutralized by a particular brand
Includes Teacher guide with sds, 30 student worksheets and guides, 8 chemplates™, 8 rubber tubes, 1 vial of universal ph indicator strips, 24 calibrated tubes with caps, 1 15ml drop control bottle of phenolphthalein solution, 8 30ml drop control
Class Size 30 Students
Product Type Antacid Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.