Learn More
Lab-Aids™ Identification of Chemical Reactions Kit

Description
Includes
Chemplates™ with spatula (12); drop-controlled bottles of solution one, Hydrochloric Acid, Sodium Carbonate, Sodium Hydroxide, Potassium Chromate, Calcium Chloride, Sodium Sulfite, Copper Sulfate and an unknown solution; worksheets and guides; and instructions with MSDS
This kit will help students hone their deductive reasoning and observation interpretation skills. Students first mix seven known solutions into any of twenty-one possible combinations, using standard lab procedures. Then observe and make notations as to how the chemicals react. Some of the chemical reactions produced by the combinations are: formation of a precipitate, color change, gas generation, exothermic reaction. After recording their observations, students must decide whether a chemical reaction has taken place, and provide evidence to support their deductions. As an extension, students can write the probable chemical equation of each reaction. Accommodates 15 groups of 2 students.
Order Info
Class size: 50 students.
Specifications
Specifications
| For Use With (Application) | To identify four types of chemical reactions, color changes, precipitates, gas generation and exothermic |
| Includes | 12 chemplates™ with spatula, drop controlled bottles of solution one, hydrochloric acid, sodium carbonate, sodium hydroxide, potassium chromate, calcium chloride, sodium sulfite, copper sulfate and an unknown solution worksheets and guid |
| Class Size | 50 Students |
| Product Type | Identification of Chemical Reactions Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.