Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab-Aids™ Properties of Acids and Bases
SCI_ED

Catalog No. S19392
Encompass_Preferred
Change view
Click to view available options
Class Size:
50 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S19392 50 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19392 Supplier Lab-Aids™ Supplier No. 82

Study, test, and observe the characteristic properties of acids and bases, prepare, collect, and test hydrogen and carbon dioxide safely, observe the effects of acids and bases on indicator dyes, measure the concentration of acids by titration of an 'unknown' and perform a neutralization experiment between an acid and a base to form a salt.

Lab-Aids™ Applied Science Concepts Kit provides a completely safe and convenient method for students to study, test, and observe the characteristic properties of acids and bases. Activities incorporate the Lab-Aids™ Chemplates. This unique lab tool eliminates the need for glassware and provides a semi-micro environment in any classroom.

Students perform a total of seven different activities: reacting metal with acids and producing hydrogen in safe quantities, collecting and safely testing hydrogen, reacting carbonates with acids and producing carbon dioxide, collecting and testing carbon dioxide, observing the effects of acids and bases on the indicator dyes, measuring the concentration of acids by titration of an 'unknown', performing a neutralization experiment.  Accommodates 3 classes, each with 12 groups of 2 students.

Specifications

For Use With (Application) Study, test, and observe the characteristic properties of acids and bases
Includes Teacher's guide with msds, 50 student worksheets and guides, 12 chemplates™ with spatula and gas collector cap, 12 gas delivery tubes, wooden splints, package, 3 test tubes, 6m hydrochloric acid, bottles, 6m acetic acid, drop control bot
Class Size 50 Students
Product Type Properties of Acids and Bases Experiment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.