Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab-Aids™ Simulated Urinalysis Kit
SCI_ED

Catalog No. S19552
Encompass_Preferred
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S19552 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19552 Supplier Lab-Aids™ Supplier No. 4

Includes

  • 15 Lab-Aids Chemplates™ with spatula
  • Three bottles each of simulated urine I, II, III, IV and V
  • Three bottles of dilute acetic acid
  • Three bottles of dilute silver nitrate
  • Three bottles of Benedict's solution
  • Five density tubes; one vial of universal indicator paper
  • 15 pH color charts
  • 30 student worksheets and guides
  • Instruction manual with Material Safety Data Sheets

Perform diagnostic lab tests. Apply analyze as a diagnostic tool.

With this kit, students analyze simulated urine samples to understand the procedures used in urinalysis and the potential applicability of urinalysis as a diagnostic tool. By adding different reagents to urine samples, students observe how each reacts with substances in the urine, providing useful evidence for identifying different substances. Students test simulated urine samples for pH, density, glucose, phosphates and chlorides. Enough reagents are provided for 30 students. A hot plate, Bunsen burner and test tubes are necessary, but not included.

Specifications

For Use With (Application) Shows the importance of urinalysis as a reflection of body health.
Includes 15 lab-aids™ chemplates™ with spatula, three bottles each of simulated urine i, ii, iii, iv and v, three bottles of dilute acetic acid, three bottles of dilute silver nitrate, three bottles of benedict's solution, five
Class Size 30 Students
Product Type Simulated Urinalysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.