Learn More
Lab Armor™ Chill Bucket™ Bag Kits
SDP
For cooling samples while keeping them dry and contamination-free without using wet ice
$434.83
Specifications
Type Chill Kit
View More Specs
Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-876-020
SDP
Encompass
View Documents
Lab Armor™
67200001
Beads, chill packs, bead bag w/ lid
Each for $434.83
Only null left
Add to Cart
10-876-021
SDP
View Documents
Lab Armor™
67200002
Beads, chill packs, bead bag N/A N/A N/A
10-876-022
SDP
Encompass
View Documents
Lab Armor™
67200005
Chill bucket, chill packs, bead bag N/A N/A N/A
Description
Description
  • Durable
  • Bead bags are made of washable mildew and rot resistant nylon mesh fabric
Chill Bucket
  • Light weight and condensation free polyurethane construction
  • Molded side knobs with lids
  • Shaped to balance in one arm
  • Can be hand washed with mild detergent or towel dried
Order Info
Additional chill packs available separately (See Cat. No. 10-876-023).
Specifications
Specifications
Chill Kit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.