Learn More
Research Products International Corp Lab Armor Insulated Mini-Tray w/ Beads
SDP
Supplier:  Research Products International Corp 299301
Encompass
The Lab Armor Insulated Mini-Tray provides a unique way to keep samples at temperature under a fume hood, on a benchtop or while being carried. The Mini-Tray is one-fourth the size of a standard ice bucket and convenient for use in tight spaces. Supplied with Lab Armor Beads and can also be used with ice. Mini-Tray is made from expanded PVC foam and has a temperature range of -196°C to +93°C.
Catalog No. 50-197-7812