Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Lab Bench Vert Organizer, 4 Pocket, Double-wide
SDP

Catalog No. 22000764
Change view
Click to view available options
Includes:
Organizer
Organizer, Pipette Aid Bracket
Organizer, Pipetter, Pipette Aid Brackets
Organizer, Pipettor Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-764 Organizer, Pipetter, Pipette Aid Brackets
22-000-788 Organizer
22-000-789 Organizer, Pipettor Bracket
22-000-772 Organizer, Pipette Aid Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-000-764 Supplier Poltex™ Supplier No. 4VTORGDBLPIPTRBRKBRK
Add to Cart
Add to Cart

Benchtop organizer for tip boxes, bottles, etc. Made of durable PETG construction.

  • Free up valuable lab bench space by going up!
  • Highly durable, 1/4 in. thick PETG construction
  • Holds pipette tips, bottles, etc.
  • Comes with optional accessories: 4 pipettor bracket and/or pipette controller bracket
  • Rubber bumpons on base for no slip placement

Specifications

Product Type Organizer
Includes Organizer, Pipetter, Pipette Aid Brackets
Width (Metric) 242 mm
For Use With (Application) Pipette Tip Boxes
Color Clear
Autoclavable Not Autoclavable
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.