Learn More
Poltex™ Lab Bench Vert Organizer, 4 Pocket, Double-wide

Description
- Free up valuable lab bench space by going up!
- Highly durable, 1/4 in. thick PETG construction
- Holds pipette tips, bottles, etc.
- Comes with optional accessories: 4 pipettor bracket and/or pipette controller bracket
- Rubber bumpons on base for no slip placement
Specifications
Specifications
| Product Type | Organizer |
| Autoclavable | Not Autoclavable |
| Width (Metric) | 242 mm |
| Color | Clear |
| Includes | Organizer, Pipette Aid Bracket |
| For Use With (Application) | Pipette Tip Boxes |
| Material | PETG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.