Learn More
Ace Glass, Inc. Lab bottle, Duran, 10L, plain, pouring ring & PTFE lined GL 45PBT cap, cs/1
SDP
Supplier:  Ace Glass, Inc. 553935
Encompass
Chemical-resistant and stable 10L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. 227 mm outer diameter x 415 mm tall. Case/1.
Catalog No. 50-978-376
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.