Learn More
New Pig Lab & Chemical Cleanup Kit
SDP
Catalog No. 18389126 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Lab & Chemical Cleanup Kit
Catalog No.
18-389-126 Type: Lab & Chemical Cleanup Kit
1 options
This item is not returnable. View return policy
Catalog No. 18-389-126 Supplier New Pig Supplier No. KIT5008
Add to Cart
Add to Cart
This item is not returnable. View return policy
Be prepared for small spills in the laboratory with our grab-and-go clean up kit for acids, bases and chemicals.
  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Easy-open, resealable bag protects kit contents from dirt and moisture
  • Single-use kit is faster and more efficient than dragging a large spill kit to a small cleanup; safer and more effective than using rags or paper towels
  • Includes hazmat absorbent pads, acid neutralizing loose absorbent and base neutralizing loose absorbent to clean up most acids, bases and chemicals
  • Disposable gloves, mini dustpan and disposal bag also included to aid cleanup
Specifications
Material Polypropylene
WARNING: Cancer - www.P65Warnings.ca.gov