Learn More
West Chester Polypropylene Lab Coats
Catalog No. 19821588 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
Cuff Style:
Elastic
Closure Type:
Snaps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style Closure Type
19-821-588 2X-Large Elastic Snaps
19-121-682D X-Large Elastic Snaps
19-121-682B Medium Elastic Snaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-821-588 Supplier West Chester Supplier No. 3512/XXL
  • $144.44
    $78.65 / Case of 30
    Save $65.79
    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard weight lab coats offer breathability and durability. Choose open wrists or elastic wrists style.
  • Lightweight protection against dry particulates
  • Made with durable polypropylene material
  • Snap front closure for secure fit and allows easy donning
  • Collared neck for better air circulation
  • Choose open or elastic wrist style
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Dust
Style Standard Length
No. of Pockets None
Latex Free Yes
Model 3512
Thickness Standard weight
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.