Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MicroBioLogics™ Lab Elite™ CRM Rhodococcus equi ATCC™ 6939™†
SDP

Catalog No. 12111318 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Rhodococcus equi
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
12-111-318 Rhodococcus equi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-111-318 Supplier MicroBioLogics™ Supplier No. 0697CRM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)

Used for quality control. Microbiologics™ Lab Elite™ CRM Rhodococcus equi ATCC™ 6939™† is self-contained device.

  • Delivered in an easy-to-use KWIK-STIK™ format
  • Traceable to reference culture
  • Highly characterized Rhodococcus equi
  • For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
  • ATCC™ 6939™†

Specifications

ATCC® 6939
Biosafety Level 2
Content And Storage 35°C
Format KWIK-STIK™
Species Rhodococcus equi

† The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.