Learn More
Ted Pella Inc LAB GRD DEWARS 350ML STANDARD
SDP
Supplier:  Ted Pella Inc 8174050
Encompass
Lab Grade Dewars 350ml Standard Base, Cylindrical Wide Mouth, Protective Covering: Mesh, 70mm id, 125mm inside depth, 86mm od, 157mm total height.
Catalog No. NC1998608
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.