Learn More
Cytiva (Formerly Pall Lab) Laboratory Manifold Coupling Device (1/pkg)
Supplier:  Cytiva 4893
Encompass_Preferred
A unique microbiology manifold that allows you to optimize testing without sacrificing cleanliness. With interchangeable components and coupling devices, the manifold takes direction from you.-Microbial cross contamination of your sample is a thing of the past. The manifold is designed to fit most laboratory autoclaves by separating into manageable components.-A larger manifold is at your reach with just a coupler. Modular design allows two manifolds to combine yielding a standard 6-place manifold which can be easily separated for disinfection and/or sterilization.-Whether you are right or left-handed, setting up your test workflow is second nature. The components interchangeable and designed with a simple slip fit to the manifold.- Manifold includes 3-place manifold base, 3 manifold valves, 1 end cap, and 1 hose barb cap. Adapters are required and sold separately This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3115
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.